Learn More
Sino Biological Mouse CPB2 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50963T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Carboxypeptidase B2 / CPB2 (Catalog#50963-M08H; Q9JHH6; Met1-Thr422). Carboxypeptidase B2 / CPB2 specific IgG was purified by Mouse Carboxypeptidase B2 / CPB2 affinity chromatography.
Catalog No. 50-237-0582