Learn More
Sino Biological Recombinant Anti-CRABP2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50535R101100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CRABP2 (rM CRABP2; Catalog#50535-M07E; P22935; Pro 2-Glu 138).
Catalog No. 50-241-2443