Learn More
Sciencell Research Laboratories MOUSE CRDC MYOCYTS FRM C57BL/6
Catalog No. NC2602432
Encompass
Isolated from postnatal day 2 or postnatal day 8 C57BL/6 mouse heart. MCM are cryopreserved at P0 and delivered frozen. Each vial contains >1 x 106 cells in 1 ml volume.
Catalog No. NC2602432 Supplier Sciencell Research Laboratories Supplier No. M620057
Isolated from postnatal day 2 or postnatal day 8 C57BL/6 mouse heart. MCM are cryopreserved at P0 and delivered frozen. Each vial contains >1 x 106 cells in 1 ml volume.