Learn More
Abcam Mouse Creatine Kinase MB ELISA Kit, 96TEST
Supplier:  Abcam AB28523196TEST
Encompass_Preferred
Mouse Creatine Kinase MB ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Creatine Kinase MB in Mouse in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5441