Learn More
American Research Products Inc Mouse Creatine Kinase MB isoen
Catalog No. 5000328492
Encompass
Mouse Creatine Kinase MB isoenzyme CK-MB ELISA Kit from CUSABIO Detection Range 1 25 mU/mL-80 mU/mL Sensitivity 0 312 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28492 Supplier American Research Products Inc Supplier No. CSBE14404M
Mouse Creatine Kinase MB isoenzyme CK-MB ELISA Kit from CUSABIO Detection Range 1 25 mU/mL-80 mU/mL Sensitivity 0 312 mU/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich