Learn More
Biomatik Corporation Mouse Creatine Kinase MB Isoenzyme (CKMB) ELISA Kit
Supplier:  Biomatik Corporation EKU03486
Encompass_Preferred
Mouse Creatine Kinase MB Isoenzyme (CKMB) ELISA Kit for detecting CKMB in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07310; Q04447. Gene ID: 12715; 12709. Target Synonyms: CK2; CK-MB; Creatine Kinase,Muscle/Brain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9030