Learn More
Biomatik Corporation Mouse Creatine Kinase, Mitochondrial 2, Sarcomeric (CKMT2) ELISA Kit
Supplier:  Biomatik Corporation EKU12336
Encompass_Preferred
Mouse Creatine Kinase, Mitochondrial 2, Sarcomeric (CKMT2) ELISA Kit for detecting CKMT2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P8J7. Gene ID: 76722. Target Synonyms: MtCK; CK-MT2; SMTCK; Mib-CK; Creatine Kinase S-type; Basic-type mitochondrial creatine kinase; Sarcomeric mitochondrial creatine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7981