Learn More
Biomatik Corporation Mouse Creatine Kinase, Mitochondrial 1A (CKMT1A) ELISA Kit
Supplier:  Biomatik Corporation EKU12334
Encompass_Preferred
Mouse Creatine Kinase, Mitochondrial 1A (CKMT1A) ELISA Kit for detecting CKMT1A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30275. Gene ID: 12716. Target Synonyms: CKMT; CKMT1-A; Mia-CK; U-MtCK; Creatine kinase U-type, mitochondrial; Ubiquitous mitochondrial creatine kinase; Acidic-type mitochondrial creatine kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7330