Learn More
Crystal Chem Inc Mouse Creatinine Assay Kit, 10-Kit Pack, 96-tests per kit
SDP
Catalog No. 501947697
Encompass_Preferred
The Mouse Creatinine Assay is used to quantify levels of creatinine in mouse serum and urine in less than 10 min. The 10-Kit Pack is convieniently provided as ten individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight of (8) µL of sample, ideal for research applications. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7697 Supplier Crystal Chem Inc Supplier No. 80356
The Mouse Creatinine Assay is used to quantify levels of creatinine in mouse serum and urine in less than 10 min. The 10-Kit Pack is convieniently provided as ten individual kits.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobinRequires only eight of (8) µL of sample, ideal for research applications. Designed for rat samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.