Learn More
Crystal Chem Inc Mouse Creatinine Assay Kit, 96-tests
SDP
Catalog No. 501947694
Encompass_Preferred
The Mouse Creatinine Assay is used to quantify levels of creatinine in mouse serum and urine in less than 10 min.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobin. Requires only eight (8) µL of sample, ideal for research applications. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7694 Supplier Crystal Chem Inc Supplier No. 80350
The Mouse Creatinine Assay is used to quantify levels of creatinine in mouse serum and urine in less than 10 min.Highlights: Fully enzymatic method with significantly reduced interference compared to alkaline picric acid. Less than 10% variance from triglyceride, ascorbic acid, bilirubin, hemoglobin. Requires only eight (8) µL of sample, ideal for research applications. Designed for mouse samples.Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.