Learn More
BIOHIPPO Mouse CRN Microsample
SDP
Catalog No. 5000314195
Encompass
Mouse CRN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CRN in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CRN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CRN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CRN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14195 Supplier BIOHIPPO Supplier No. BHE15212513
Mouse CRN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CRN in Serum plasma and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 058 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CRN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CRN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CRN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color