Learn More
American Research Products Inc Mouse cross linked C telopepti
Catalog No. 5000329033
Encompass
Mouse cross linked C-telopeptide of type I collagen (CTX- ) ELISA Kit from CUSABIO Detection Range 0 23 ng/ml-15 ng/ml Sensitivity 0 12 ng/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-29033 Supplier American Research Products Inc Supplier No. CSBE12782M
Mouse cross linked C-telopeptide of type I collagen (CTX- ) ELISA Kit from CUSABIO Detection Range 0 23 ng/ml-15 ng/ml Sensitivity 0 12 ng/ml Sample serum plasma tissue homogenates Method Competitive