Learn More
Biomatik Corporation Mouse Cross Linked C-Telopeptide Of Type I Collagen (CTXI) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12147
Encompass_Preferred
Mouse Cross Linked C-Telopeptide Of Type I Collagen (CTXI) Mini sample ELISA Kit for detecting CTXI in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 51.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11087; Q01149. Gene ID: 12842; 12843. Target Synonyms: CTX-I; CTX1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5386