Learn More
Raybiotech Inc MOUSE CRP ELISA
Supplier:  Raybiotech Inc ELMCRP1
Encompass_Preferred
RayBio Mouse CRP (C-Reactive Protein) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 0.15 ng/ml - 40 ng/ml
Catalog No. 50-104-4619