Learn More
BIOHIPPO Mouse CRP Microsample
SDP
Catalog No. 5000315261
Encompass
Mouse CRP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CRP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 56-100 g/mL with a sensitivity of 1 09 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CRP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CRP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CRP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15261 Supplier BIOHIPPO Supplier No. BHE15212680
Mouse CRP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CRP in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 56-100 g/mL with a sensitivity of 1 09 g/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Mouse CRP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Mouse CRP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Mouse CRP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color