Learn More
Sino Biological Anti-C-Reactive Protein Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50409RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CRP (rM CRP; Catalog#50409-M08H; NP_031794.3; Met 1-Ser 225). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8435