Learn More
Biomatik Corporation Mouse Crystallin Gamma S (CRYgS) ELISA Kit
Supplier:  Biomatik Corporation EKU08346
Encompass_Preferred
Mouse Crystallin Gamma S (CRYgS) ELISA Kit for detecting CRYgS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O35486. Gene ID: 12970. Target Synonyms: CRYG8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4779