Learn More
Sino Biological Anti-M-CSF/CSF1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51112T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse M-CSF / CSF-1 (Catalog#51112-M08H; P07141-1; Met1-Glu262). M-CSF / CSF-1 specific IgG was purified by Mouse M-CSF / CSF-1 affinity chromatography.
Catalog No. 50-240-8376