Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50059T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CSF1R (rM CSF1R; Catalog#50059-M08H; NP_001032948.2; Met1-Ser511). CSF1R specific IgG was purified by Mouse CSF1R affinity chromatography.
Catalog No. 50-240-8163