Learn More
Sino Biological Anti-M-CSFR/CSF1R Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50059RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CSF1R / MCSF Receptor / CD115 (rM CSF1R / MCSF Receptor / CD115; Catalog#50059-M08H; NP_001032948.2; Met 1-Ser 511). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1848