Learn More
Sino Biological Recombinant Mouse M-CSFR/CSF1R Protein (ECD, hFc Tag)
Supplier:  Sino Biological 50059M02H100
Encompass_Preferred
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) (Met1-Ser511) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-3507