Learn More
Sino Biological Recombinant Mouse M-CSFR/CSF1R Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50059M08HB100
Encompass_Preferred
A DNA sequence encoding the mouse CSF1R (NP_001032948.2) extracellular domain (Met1-Ser511) was fused with a polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vitro.
Catalog No. 50-255-3506