Learn More
Sino Biological Mouse CSF2 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371109
Encompass_Preferred
Each vial contains 11 ng of recombinant Mouse GM-CSF/CSF2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 120 pg/mL is recommended.
Catalog No. 50-237-1109 Supplier Sino Biological Supplier No. SEK5104815
Each vial contains 11 ng of recombinant Mouse GM-CSF/CSF2. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 120 pg/mL is recommended.