Learn More
Sino Biological Recombinant Anti-GM-CSF/CSF2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 51048R017100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse GM-CSF/CSF2 (rM GM-CSF/CSF2; Catalog#51048-M07H; P01587; Ala18-Lys141).
Catalog No. 50-241-0071