Learn More
Sino Biological Anti-CK2 alpha/CSNK2A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50818RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CSNK2A1 / CK2A1 (rM CSNK2A1 / CK2A1; Catalog#50818-M20B; Q60737; Met1-Gln391). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3071