Learn More
Sino Biological Mouse CST7 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50236T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cystatin 7 / CST7 (Catalog#50236-M02H; O89098; Met1-Gln144). Cystatin 7 / CST7 specific IgG was purified by Mouse Cystatin 7 / CST7 affinity chromatography.
Catalog No. 50-237-3778