Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CTACK (CCL27) Recombinant Protein, PeproTech®

Catalog No. 2502620UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
250-26-20UG 20 μg
25026100UG 100 μg
250-26-1MG 1 mg
25026250UG 250 μg
25026500UG 500 μg
250-26-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 250-26-20UG Supplier Gibco™ Supplier No. 2502620UG
  • $225.00
    $168.65 / Each of 1

    Save $56.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine CTACK is a 10.9 kDa protein containing 95 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CCL27 gene is one of several CC cytokine genes clustered on the p-arm of chromosome 9. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteines. The protein encoded by this gene is chemotactic for skin-associated memory T lymphocytes. This cytokine may also play a role in mediating homing of lymphocytes to cutaneous sites. It specifically binds to chemokine receptor 10 (CCR10). Studies of a similar murine protein indicate that these protein-receptor interactions have a pivotal role in T cell-mediated skin inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z1X0
For Use With (Application) Control, Bioactivity
Formulation Protein with no preservative
Gene ID (Entrez) 20301
Molecular Weight (g/mol) 10.9 kDa
Name Mouse CTACK (CCL27)
Purification Method Purified
Quantity 20 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias ALP; AW558992; CC chemokine ILC; C-C motif chemokine 27; C-C motif chemokine ligand 27; Ccl27; Ccl27a; chemokine (C-C motif) ligand 27; chemokine (C-C motif) ligand 27A; CTACK; CTAK; cutaneous T-cell attracting chemokine; cutaneous T-cell-attracting chemokine; ESkine; IL-11 R-alpha-locus chemokine; IL-11 Ralpha-locus chemokine; ILC; PESKY; SCYA27; Scya27a; Skinkine; skinskine; small inducible cytokine A27; small inducible cytokine A27a; small inducible cytokine subfamily A (Cys-Cys), member 27; small-inducible cytokine A27
Common Name CCL27
Gene Symbol Ccl27a
Biological Activity Determined by its ability to chemoattract human peripheral blood lymphocytes using a concentration range of 10.0-100.0 ng/ml
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence LPLPSSTSCC TQLYRQPLPS RLLRRIVHME LQEADGDCHL QAVVLHLARR SVCVHPQNRS LARWLERQGK RLQGTVPSLN LVLQKKMYSN PQQQN
Content And Storage -20°C
Activity Determined by its ability to chemoattract human peripheral blood lymphocytes using a concentration range of 10.0-100.0 ng/ml.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.