Learn More
Abcam Mouse CTGF ELISA Kit, 96TEST
Supplier:  Abcam AB28983896TEST
Encompass_Preferred
Mouse CTGF ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CTGF in Mouse in Plasma, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2590274