Learn More
Sino Biological Anti-CTLA-4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50503RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CTLA-4 / CD152 (rM CTLA-4 / CD152; Catalog#50503-M08H; NP_033973.2; Met 1-Phe 162). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1309