Learn More
Sino Biological Recombinant Mouse CTLA-4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 50503M08HB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of mouse CTLA4 (NP_033973.2) (Met1-Phe162) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3244