Learn More
Sino Biological Recombinant Mouse CTLA-4 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 50503M49HB100
Encompass_Preferred
A DNA sequence encoding the Mouse CTLA4 (NP_033973.2)(Met1-Phe162) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0248