Learn More
Life Diagnostics Inc Ultra-Sensitive Mouse Cardiac Troponin-I ELISA, 96 wells, serum/plasma samples, sandwich ELISA, colorimetric detection, 0.039 - 2.5 ng/ml standard curve, 3 hours 20 minutes incubation
SDP
Supplier:  Life Diagnostics Inc CTNI1US
Encompass_Preferred
Ultra-Sensitive Mouse Cardiac Troponin-I ELISA, 96 wells, serum/plasma samples, sandwich ELISA, colorimetric detection, 0.039 - 2.5 ng/ml standard curve, 3 hours 20 minutes incubation
Catalog No. NC1052736