Learn More
Biomatik Corporation Mouse cTnT/TNNT2 (Troponin T Type 2, Cardiac) ELISA Kit
Supplier:  Biomatik Corporation EKF58659
Encompass_Preferred
Mouse cTnT/TNNT2 (Troponin T Type 2, Cardiac) ELISA Kit for detecting cTnT; TNNT2 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P50752. Target Synonyms: cTnT; TNNT2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9167