Learn More
Sino Biological Anti-Cathepsin B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50084T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cathepsin B / CTSB (rM Cathepsin B / CTSB; Catalog#50084-M08H; P10605; Met1-Phe339). Cathepsin B / CTSB specific IgG was purified by Mouse Cathepsin B / CTSB affinity chromatography.
Catalog No. 50-241-0283