Learn More
Sino Biological Recombinant Anti-Cathepsin D Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50127R102100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Cathepsin D / CTSD (rM Cathepsin D / CTSD; Catalog#50127-M08H; NP_034113.1; Met 1-Leu 410).
Catalog No. 50-255-3447