Learn More
Sino Biological Anti-Cathepsin D Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50127RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CTSD (rM CTSD; Catalog#50127-M08H; NP_034113.1; Met 1-Leu 410). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3446