Learn More
Sino Biological Anti-Cathepsin L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50015T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cathepsin L/CTSL (rM Cathepsin L/CTSL; Catalog#50015-M08H; P06797; Met1-Asn334). Cathepsin L/CTSL specific IgG was purified by Mouse Cathepsin L/CTSL affinity chromatography.
Catalog No. 50-241-2568