Learn More
Sino Biological Anti-Cathepsin L Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50015RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Cathepsin L / CTSL1 (Catalog#50015-M08H; P06797; Met1-Asn334). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8315