Learn More
Biomatik Corporation Mouse CUB And Zona Pellucida Like Domains Protein 1 (CUZD1) ELISA Kit
Supplier:  Biomatik Corporation EKU10096
Encompass_Preferred
Mouse CUB And Zona Pellucida Like Domains Protein 1 (CUZD1) ELISA Kit for detecting CUZD1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.068ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P70412. Gene ID: 16433. Target Synonyms: ERG-1; UO-44; Transmembrane protein UO-44. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8373