Learn More
Biomatik Corporation Mouse Cubilin (CUBN) ELISA Kit
Supplier:  Biomatik Corporation EKU12198
Encompass_Preferred
Mouse Cubilin (CUBN) ELISA Kit for detecting CUBN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9JLB4. Gene ID: 65969. Target Synonyms: IFCR; MGA1; gp280; Intrinsic Factor Cobalamin Receptor; 460 kDa receptor; Intestinal intrinsic factor receptor; Intrinsic factor-vitamin B12 receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8010