Learn More
Sino Biological Anti-CXADR/CAR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50019RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXADR (rM CXADR; Catalog#50019-M08H; NP_001020363.1; Met 1-Gly 237). CXADR specific IgG was purified by Mouse CXADR affinity chromatography.
Catalog No. 50-241-2555