Learn More
American Research Products Inc Mouse CXC chemokine ligand 16
Catalog No. 5000329231
Encompass
Mouse CXC-chemokine ligand 16 CXCL16 ELISA Kit from CUSABIO Detection Range 7 8125 pg/mL-500 pg/mL Sensitivity 3 012 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29231 Supplier American Research Products Inc Supplier No. CSBE08873M
Mouse CXC-chemokine ligand 16 CXCL16 ELISA Kit from CUSABIO Detection Range 7 8125 pg/mL-500 pg/mL Sensitivity 3 012 pg/mL Sample serum plasma tissue homogenates Method Sandwich