Learn More
Sino Biological Anti-SDF-1/CXCL12 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50025T48100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL12 / SDF-1 beta (rM CXCL12 / SDF-1 beta; Catalog#50025-MNAE; P40224-1; Lys22-Lys89). CXCL12 / SDF-1 beta specific IgG was purified by Mouse CXCL12 / SDF-1 beta affinity chromatography.
Catalog No. 50-241-2541