Learn More
Sino Biological Recombinant Mouse SDF-1/CXCL12 Protein
Supplier:  Sino Biological 50025MNAE1
Encompass_Preferred
A DNA sequence encoding the mature form of mouse CXCL12 (P40224-1) (Lys22-Lys89) was expressed and purified with an initial Met.
Catalog No. 50-255-3548