Learn More
Sino Biological Anti-CXCL16 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50142T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL16 / SR-PSOX (rM CXCL16 / SR-PSOX; Catalog#50142-M08H; NP_075647.3; Met1-Trp201). CXCL16 / SR-PSOX specific IgG was purified by Mouse CXCL16 / SR-PSOX affinity chromatography.
Catalog No. 50-255-3415