Learn More
Sino Biological Anti-CXCL16 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50142RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL16 / SR-PSOX (Catalog#50142-M08H; NP_075647.3; Met1-Trp201). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-3416