Learn More
Sino Biological Recombinant Mouse CXCL16 Protein (His Tag)
Supplier:  Sino Biological 50142M08H1
Encompass_Preferred
A DNA sequence encoding the mouse CXCL16 (NP_075647.3) extracellular domain (Met 1-Trp 201) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-255-3417