Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse CXCL16 Recombinant Protein, PeproTech®

Catalog No. p-7245772
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine CXCL16 is a 9.9 kDa protein containing 88 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

CXCL16 is a member of the CXC chemokine family and signals through the CXCR6 receptor. CXCL16 may play a role in attracting lymphocyte subsets during inflammation and may facilitate certain immune responses. The chemokine domain of CXCL16 contains six cysteine residues, including the four highly conserved cysteine residues characteristic of CXC chemokines. The CXCL16 gene codes for a 273 amino acid polypeptide, which includes a 29 amino acid cytoplasmic domain and transmembrane sequence containing approximately 20 amino acids. The extracellular portion of CXCL16 contains a chemokines domain and an extended C-terminal mucin-like stalk sequence. The extracellular domain contains 89 amino acid residues (86 a.a. residues for the murine homolog).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8BSU2
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 66102
Molecular Weight (g/mol) 9.9 kDa
Name Mouse CXCL16
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 0910001K24Rik; AV290116; b2b498Clo; BB024863; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 16; CXC; Cxc chemokine ligand 16; CXC motif chemokine; C-X-C motif chemokine 16; C-X-C motif chemokine ligand 16; CXCL; Cxcl16; CXCL16v1; CXCL16v2; CXCLG16; Mutant line 498; scavenger receptor for phosphatidylserine and oxidized low density lipoprotein; SCYB16; Small-inducible cytokine B16; Srpsox; SR-PSOX; SR-PSOX/CXCL16; Transmembrane chemokine CXCL16; UNQ2759/PRO6714; zinc finger, MYND-type containing 15; Zmynd15
Common Name CXCL16
Gene Symbol Cxcl16
Biological Activity Determined by its ability to chemoattract murine lymphocytes using a concentration of 20-1000 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence NQGSVAGSCS CDRTISSGTQ IPQGTLDHIR KYLKAFHRCP FFIRFQLQSK SVCGGSQDQW VRELVDCFER KECGTGHGKS FHHQKHLP
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.