Learn More
Sino Biological Recombinant Anti-GRO Beta/CXCL2 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 50070R019100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse GRO beta/CXCL2 (rM GRO beta/CXCL2; Catalog#50070-M28E; P10889-1; Ala28-Asn100).
Catalog No. 50-240-8552