Learn More
Sino Biological Anti-GRO Beta/CXCL2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50070T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse CXCL2 / MIP-2 (rM CXCL2 / MIP-2; Catalog#50070-M28E; P10889-1; Ala28-Asn100). CXCL2 / MIP-2 specific IgG was purified by Mouse CXCL2 / MIP-2 affinity chromatography.
Catalog No. 50-241-1341